Flame-retardant high visibility fleece jacket, Rainier 496Z
Supplier: SIOEN
Certificates
About this item
Flame retardant safety fleece jacket, made of 100% flame retardant polyester fleece, double-sided, with fixed flame retardant cotton lining in front.
- Soft fleece fabric and good insulation from cold
- Straight collar, hidden zip closure
- Two inset pockets with zip closure
- Elastic at end of sleeves
- Elastic drawstring in hem
- Stitched seams
Back length: 77 cm (L)
Weight: 275 g/m²
Certifications: EN ISO 14116 (Index 1/12H/40), EN ISO 20471 (Class 2), EN 14058 (Class 1XXXX)
Specifications
- Catalog No:
- SIOE496Z/279/M
- SIOE496Z/279/L
- SIOE496Z/279/3XL
- SIOE496Z/279/2XL
- SIOE496Z/279/1XL
- SIOE496Z/278/S
- SIOE496Z/278/M
- SIOE496Z/278/L
- SIOE496Z/278/3XL
- SIOE496Z/278/2XL
- SIOE496Z/278/1XL
- SIOE496Z/279/S
- Modell:
- Rainer 496Z
- Rainer 496Z
- Rainer 496Z
- Rainer 496Z
- Rainer 496Z
- Rainer 496Z
- Rainer 496Z
- Rainer 496Z
- Rainer 496Z
- Rainer 496Z
- Rainer 496Z
- Rainer 496Z
- Outfit applikationer:
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Risk area:
- Flame-retardant, Cold
- Flame-retardant, Cold
- Flame-retardant, Cold
- Flame-retardant, Cold
- Flame-retardant, Cold
- Flame-retardant, Cold
- Flame-retardant, Cold
- Flame-retardant, Cold
- Flame-retardant, Cold
- Flame-retardant, Cold
- Flame-retardant, Cold
- Flame-retardant, Cold
- Kön:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Stängningsstorlek:
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Tyg:
- 100% flame retardant polyester fleece
- 100% flame retardant polyester fleece
- 100% flame retardant polyester fleece
- 100% flame retardant polyester fleece
- 100% flame retardant polyester fleece
- 100% flame retardant polyester fleece
- 100% flame retardant polyester fleece
- 100% flame retardant polyester fleece
- 100% flame retardant polyester fleece
- 100% flame retardant polyester fleece
- 100% flame retardant polyester fleece
- 100% flame retardant polyester fleece
- Vikt:
- 275 g/m²
- 275 g/m²
- 275 g/m²
- 275 g/m²
- 275 g/m²
- 275 g/m²
- 275 g/m²
- 275 g/m²
- 275 g/m²
- 275 g/m²
- 275 g/m²
- 275 g/m²