Order Entry
Sweden
ContactUsLinkComponent
Bromofenolblått 0,01% i aceton, VOLUSOL®
Bromofenolblått 0,01% i aceton, VOLUSOL®
Katalog # VWRS86014.330
Supplier:  VWR Collection
CAS Number:  
Bromofenolblått 0,01% i aceton, VOLUSOL®
Katalog # VWRS86014.330
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Förpackningstyp:
    Plastic drum
  • Förp.:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :669.97 g/mol
Storage Temperature:Omgivande
EINECS:204-086-2
UN:1090
ADR:3,II
 :GHS02
 :GHS07
CAS Number:115-39-9
Formula:C₁₉H₁₀Br₄O₅S
MDL Number:MFCD00005875