High visibility winter rain jacket, Stormflash 2006
: SIOEN
These water- and windproof jackets are made of Ultratex fabric: 100 % polyester fabric with 100% PU coating. With fixed quilted lining and knitted wind cuffs in sleeves.
- Straight collar with foldaway hood
- Zip closure under flap with press studs
- Two inset chest pockets
- Two patched pockets with flap closed by press studs
- Kimono sleeves
- Underarm ventilation
- Taped seams
Back length: 90 cm
Weight: 200 g/m²
: EN 471 (3-2), ENV 343 (3-1), ENV 342 (Class 3-1 / Icl,r=0,274 m² K/W), EN ISO 13688
- Catalog No:
- SIOE2006/278/2XL
- SIOE2006/279/S
- SIOE2006/279/L
- SIOE2006/279/3XL
- SIOE2006/279/2XL
- SIOE2006/279/1XL
- SIOE2006/278/S
- SIOE2006/278/M
- SIOE2006/278/L
- SIOE2006/278/3XL
- SIOE2006/278/1XL
- SIOE2006/279/M
- Modell:
- 2006 Stormflash
- 2006 Stormflash
- 2006 Stormflash
- 2006 Stormflash
- 2006 Stormflash
- 2006 Stormflash
- 2006 Stormflash
- 2006 Stormflash
- 2006 Stormflash
- 2006 Stormflash
- 2006 Stormflash
- 2006 Stormflash
- Outfit applikationer:
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Risk area:
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Kön:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Stängningsstorlek:
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Tyg:
- 100% polyester fabric with 100% PU coating
- 100% polyester fabric with 100% PU coating
- 100% polyester fabric with 100% PU coating
- 100% polyester fabric with 100% PU coating
- 100% polyester fabric with 100% PU coating
- 100% polyester fabric with 100% PU coating
- 100% polyester fabric with 100% PU coating
- 100% polyester fabric with 100% PU coating
- 100% polyester fabric with 100% PU coating
- 100% polyester fabric with 100% PU coating
- 100% polyester fabric with 100% PU coating
- 100% polyester fabric with 100% PU coating
- Vikt:
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²
- 200 g/m²