Lauryl tryptose broth tubes
HACH2162315
: Hach
:
- Förp.:15
- Standard:-
- Beskrivning:Lauryl tryptose broth tubes
Lauryl tryptose broth tubes, single strength, selectivity: total coliform bacteria (presumptive).
- Save time and labour costs with ready to use MPN tubes, with minimal training requirements