Lauryl tryptose broth MPN tubes, concentrated
Katalog # HACH2101415
Leverantör: Hach
CAS-nummer:
Specifikationer
- Förp.:15
- Standard:-
- Beskrivning:Lauryl tryptose broth MPN tubes, concentrated
Specifikationer
Om denna produkt
Lauryl tryptose broth MPN tubes, concentrated. Selectivity: total coliform bacteria (presumptive).
- Save time and labour costs with ready to use MPN tubes, with minimal training requirements