Micro-tissue grinders
Type G 3 with pestle, 0,1 - 1 ml, ~8×50 mm
Katalog # ROTH0298.1
Marketsource
Leverantör: Roth Carl
Gå till:
Product Details & Documents
These micro-tissue grinders with pestles made of borosilicate glass are for pulverising and homogenising minute quantities.
Specifikationer
- Innerdiam.×H:~8×50 mm
- Kapacitet:0,1 - 1 ml
- Beskrivning:Type G 3 with pestle