Capillary columns, CP-Select CB for MTBE
Katalog # VARICP7528
Leverantör: VARIAN
Specifikationer
- Kolumnformat:Capillary
- Skikttjocklek:0,25 µm
- Ø inr.:0,25 mm
- Längd:50 m
- Phase:CP-Select CB for MTBE
- Förp.:1
Specifikationer
Om denna produkt
Ideal as primary or confirmation column.
- Engineered for analysis of MTBE in reformulated gasoline
- Unique selectivity for MTBE
- Broad dynamic range for quantification of MTBE