Orders LinkContactUsLinkComponent
Sedimentation cones, Imhoff type, Azlon®
Sedimentation cones, Imhoff type, Azlon®
Imhoff cones 1000 ml, bottom end with stopper
Katalog # AZLOSWX008
Leverantör:   SciLabware
Sedimentation cones, Imhoff type, Azlon®
Katalog # AZLOSWX008
Leverantör:  SciLabware
Sedimentation cones, Imhoff type, Azlon®
Sedimentation cones, Imhoff type, Azlon®
Imhoff cones 1000 ml, bottom end with stopper
Price
895,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
Colourless cone, made up of transparent styrene-acrylonitrile S.A.N polymer.
  • With moulded graduations
  • Stopper at the bottom facilitates the removal of sample and cleaning
Certifications: Complies to DIN 12672
Specifikationer
  • Typ:
    bottom end with stopper
  • Beskrivning:
    Imhoff cones 1000 ml
Accessories
Produktinformation
  • Katalog #
    Tillgänglighet
    Price
  • For 2 cones
    For 2 cones
    AZLOSWX004
    Log in för att kolla tillgänglighet och ert pris
    1 * 1 Items - 4 925,00 kr