Büchner funnels, PYREX®
Catalog # CORN36060-30C
Supplier: Corning
Specifications
- Platt Ø:30 mm
- Porositet:Coarse
- Kapacitet:30 ml
- Seq:03
- Förp.:1
Specifications
About this item
PYREX®, borosilicate glass, with sintered glass disc.
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
- Very good temperature resistance
Especially useful where filter paper would be attacked by material being filtered. Top edge is beaded.
Caution: Disc should not be subjected to pressure differential in excess of 1,01 bar.