Cut resistant gloves, KryTech 395
Supplier: MAPA
Certificates
About this item
These multi-purpose nitrile gloves with textile lining combine protection against cuts, liquids and contact heat up to 250 °C.
- Combined long lasting chemical protection and cut protection
- Entirely liquid-proof, withstanding mechanical aggression
- Proven long-lasting performance, measured after 5 washes
- Good grip for picking up wet and slippery objects safely
Ideal for sheet metal handling, metal rolling, stamping and assembly, cutting aluminium strips, as well as collection and sorting of industrial and household waste.
Certifications: EN 388:2016 (4X43D), EN ISO 374-1 Type B (JKOPT), EN 374-5, EN 407 (X1XXXX), ISO 13997: 20,4N (ANSI Level 3, 2080 gg); Cat. III
Specifications
- Catalog No:
- MAPA34395328
- MAPA34395320
- MAPA34395329
- Modell:
- KryTech 395
- KryTech 395
- KryTech 395
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Fodermaterial:
- Bomull
- Bomull
- Bomull
- Kragmodell:
- Rak
- Rak
- Rak
- EN standard:
- EN 388:2016 (4X43D), EN ISO 374-1 Type B (JKOPT), EN 374-5, EN 407 (X1XXXX)
- EN 388:2016 (4X43D), EN ISO 374-1 Type B (JKOPT), EN 374-5, EN 407 (X1XXXX)
- EN 388:2016 (4X43D), EN ISO 374-1 Type B (JKOPT), EN 374-5, EN 407 (X1XXXX)