Heat and cold resistant gloves, TempTec 332
Supplier: MAPA
Certificates
About this item
These neoprene gloves with knitted thermal protection provides good thermal insulation and resistance to a wide range of chemicals.
- Multi-purpose chemical resistance (acids, aliphatic solvents) increased by the thickness of the material
- Excellent thermal insulation (against heat and cold), due to double-knit cotton
- Good grip of wet or slippery objects, due to embossed texture
Ideal for a variety of industrial applications including chemical treatments of metals, oil quenching of steel, boiler room work, handling of hot piping, handling of moulds in the ceramic industry, road works in cold conditions, steam cleaning and sampling of hot petrochemical products.
Certifications: EN 388:2016 (2212X), EN ISO 374-1 Type A (ACLMNS), EN 407 (X2XXXX), EN 511 (111); Cat. III
Specifications
- Catalog No:
- 112-5558
- 112-5557
- 112-5556
- Modell:
- 332
- 332
- 332
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Fodermaterial:
- Bomull
- Bomull
- Bomull
- Yta:
- Pebbled
- Pebbled
- Pebbled
- Kragmodell:
- Rak
- Rak
- Rak
- EN standard:
- EN 388:2016 (2212X), EN ISO 374-1 Type A (ACLMNS), EN 407 (X2XXXX), EN 511 (111)
- EN 388:2016 (2212X), EN ISO 374-1 Type A (ACLMNS), EN 407 (X2XXXX), EN 511 (111)
- EN 388:2016 (2212X), EN ISO 374-1 Type A (ACLMNS), EN 407 (X2XXXX), EN 511 (111)