Cesium chloride-lanthanum chloride buffer solution acc. to Schinkel for atomic absorption spectroscopy (AAS), Supelco®
Catalog # 1.16755.1000
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Plastic bottle
- Förp.:1 L
- Beskrivning:Cesium chloride-lanthanum chloride buffer solution acc. to Schinkel
- M-Clarity quality level:MQ100
Specifications
About this item
10 g/L CsCl + 100 g/L La in dilute hydrochloric acid.