Racks for Stomacher® blender bags
Katalog # 432-3581
Leverantör: DELTALAB
Specifikationer
- Fack:14
- Beskrivning:Rack for 14 Stomacher® bags, stainless steel
- B×D×H:390×200×240 mm
- Förp.:1
Specifikationer
Om denna produkt
These racks are made of stainless steel wire with side handles incorporated.
- Very good chemical resistance
- Very good temperature resistance
- Sturdy