Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Flaskor
Dunkar
Carboys, narrow neck, LDPE, with tubulation
Carboys, narrow neck, LDPE, with tubulation
Carboys, narrow neck, LDPE, with tubulation
Leverantör:
SEMADENI
Certifikat
Expandera bilden
Carboys, narrow neck, LDPE, with tubulation
Leverantör:
SEMADENI
Om denna produkt
LDPE, translucent.
Good chemical resistance
Good temperature resistance
With silicone tubing and pinchcock