ContactUsLinkComponent
Sieve homogenisers, Cellector™
Sieve homogenisers, Cellector™
Katalog # BELC1985-85000
Leverantör:  BELLCO GLASS
Sieve homogenisers, Cellector™
Katalog # BELC1985-85000
Leverantör:  BELLCO GLASS
Specifikationer
  • Beskrivning:
    Cellector™, complete, 85 ml (pan)
  • Seq:
    01
  • Förp.:
    1
Specifikationer
Om denna produkt
Cellector™ homogenisers, made of stainless steel, are for preparation of cell suspensions and gel extraction of proteins.
Delivery: Cellector™ complete includes a pan, glass pestle, key and nine screens (one each) 10 to 150 mesh size.