Surgical gloves, latex, sterile, Gammex® Latex Moisturizing
Leverantör: ANSELL MEDICAL PRODUCTS
Certifikat
Om denna produkt
These disposable gloves integrate the new HydraSoft® technology into their inner coating and therefore rehydrate your skin as you wear them. The HydraSoft® technology from Ansell is designed to retain moisture and helps keep your skin moist and supple to fulfil its natural barrier function.
- Powder-free, micro-textured finish
- HydraSoft® is gentle to sensitive skin, yet provides robust protection against drying or cracking
- Donning and removal of the gloves is fast and easy, even with damp hands
- Micro-roughened outer surface that provides a good grip on your instruments, whether operating conditions are wet or dry
- Very high material elasticity and flexibility, ensuring you have the tactile feedback needed to adequately judge the pressure to be applied
- Very low latex protein and chemical accelerator residue content
- Skin-friendly formulation, free of thiurams and MBT (mercaptobenzothiazoles)
Certifications: EN 455 (1-4), EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS; AQL 0,65
Specifikationer
- Catalog No:
- ANSM330049065
- ANSM330049070
- ANSM330049095
- ANSM330049080
- ANSM330049085
- ANSM330049055
- ANSM330049090
- Modell:
- Latex Moisturizing
- Latex Moisturizing
- Latex Moisturizing
- Latex Moisturizing
- Latex Moisturizing
- Latex Moisturizing
- Latex Moisturizing
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Latex/latexfri:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Pudrad/puderfri:
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Yta:
- Micro-textured
- Micro-textured
- Micro-textured
- Micro-textured
- Micro-textured
- Micro-textured
- Micro-textured
- Kragmodell:
- Rak
- Rak
- Rak
- Rak
- Rak
- Rak
- Rak
- EN standard:
- EN 455 (1-4), EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
- EN 455 (1-4), EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
- EN 455 (1-4), EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
- EN 455 (1-4), EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
- EN 455 (1-4), EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
- EN 455 (1-4), EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
- EN 455 (1-4), EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS