Surgical gloves, latex, sterile, Medi-Grip® PF
Katalog # ANSM330206085
Leverantör: ANSELL MEDICAL PRODUCTS
Specifikationer
- Handskstorlek:8,5
- Förp.:50 pairs/box
- Ej handspecifika/handspecifika:Handspecifika
- Engångs/Återanvändbar:Engångs
- Pudrad/puderfri:Puderfria
- Färg:Ofärgad
- Längd:285 mm
- Seq:08
- EN standard:EN 455
- Kragmodell:Rak
- Yta:Micro-textured
Specifikationer
Om denna produkt
This glove has a very low protein level: only <50 μg/g (Modified Lowry Method - EN 455-3). This significantly reduces the potential for type I allergies. Suitable for surgical procedures.
- Powder-free, micro-textured finish
- Chlorination treatment makes donning easy
- Straight and tapered cuff provides a secure hold on the gown
- Special latex formulation makes the glove soft and comfortable, eliminating hand fatigue
- Micro-textured surface is designed to ensure a secure grip on instruments
Certifications: EN 455; AQL 0,65