ContactUsLinkComponent
Foot sprays, Stokoderm® Foot Care
Foot sprays, Stokoderm® Foot Care
Catalog # 115-2354
Supplier:  Deb-STOKO
undefined
Foot sprays, Stokoderm® Foot Care
Catalog # 115-2354
Supplier:  Deb-STOKO
Specifications
  • Beskrivning:
    Foot spray, Stokoderm® Foot Care, 100 ml bottle
  • Förp.:
    25
Specifications
About this item
This foot spray helps to protect feet by preventing excessive perspiration, for instance when wearing closed in footwear, such as safety shoes, rubber boots etc. The alcoholic solution contains fragrances of essential oils and Hamamelis extract, which strengthens the skin and prevents softening.
  • Alcohol base provides refreshing effect and helps provide effective protection against fungal skin infections
  • Essential oils provide protection against any unpleasant smells that may arise from the bacterial breakdown of sweat
  • Convenient spray form: Easy to use and apply to all areas
  • Also suitable for deodorising footwear
Caution: *Produkten/produkterna på denna sida ligger utanför tillämpningsområdet för EU -direktivet 2019/904