Quartz cuvettes for spectrophotometers
Katalog # WAPR80-2103-68
Leverantör: Biochrom
Specifikationer
- Skiktdjup:5 mm
- Typ:Ultra micro
- Volym:5 – 7 µl
- Beskrivning:Quartz cuvettes
- Seq:010
- Förp.:1
Specifikationer
Om denna produkt
Life science cuvettes for methods with rapid scanning, such as nucleic acid quantification, proteins and cell density.