Magnetic stirring bars, micro
Leverantör: SciLabware
Certifikat
Om denna produkt
PTFE covered, suitable for use in small vessels, e.g. test tubes and vials.
- Very good chemical resistance
- Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)