Digital refractometers, PR-α series
Leverantör: Atago
Certifikat
Om denna produkt
Modern, attractive refractometers for sugar applications.
- Different colours to identify the models
- Simple two-button operation
- Calibration with water
External light interference (ELI) to take accurate measurements in daylight or near a window.
Delivery: PR-301α is supplied with a calibration standard (7 ml) and round sheets.
Specifikationer
- Catalog No:
- 635-0150
- 635-0151
- 635-0152
- 635-0149
- Område för temperaturkompensation:
- 5/10...40 °C
- 5/10...40 °C
- 5/10...40 °C
- 5/10...40 °C
- Elektrisk:
- 9 V 006p dry battery
- 9 V 006p dry battery
- 9 V 006p dry battery
- 9 V 006p dry battery
- W×D×H:
- 170×90×40 mm
- 170×90×40 mm
- 170×90×40 mm
- 170×90×40 mm
- Vikt:
- 300 g
- 300 g
- 300 g
- 300 g