ContactUsLinkComponent
Brilliant cresyl blue ALD, certified by the Biological Stain Commission, Sigma-Aldrich®
Brilliant cresyl blue ALD, certified by the Biological Stain Commission, Sigma-Aldrich®
Katalog # B5388-10G
CAS-nummer:  
undefined
Brilliant cresyl blue ALD, certified by the Biological Stain Commission, Sigma-Aldrich®
Katalog # B5388-10G
CAS-nummer:  
Specifikationer
  • Förpackningstyp:
    Glass bottle for solids
  • Förp.:
    10 g
  • Seq:
    3236
Specifikationer
Specifikationstestresultat
  • Dye content:
    ≥60%
Specifikationstestresultat
Om denna produkt
 :
 :801.96 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:279-675-0
CAS-nummer:51716-96-2
Formel:C₃₄H₄₂Cl₄N₈O₂Zn
MDL-nummer:MFCD00148901