VWR® Q Path® Slide Cabinets
Catalog # 631-1064
Supplier: VWR Collection
Specifications
- Antal objektglas:1860
- Färg:Gul
- Beskrivning:Slide cabinet
- Material:Kartong
- B×D×H:290×295×90 mm
- Förp.:10
Specifications
About this item
Slide cabinet for long-term storage.
- Rigid
- Laminated
- Stackable
W×D×H: 290×295×90 mm
Colour: Yellow