Dirt and stain remover, Miraclean
129-0991
- Färg:Vit
- Beskrivning:Miraclean dirt remover
- L×B:120×75 mm
- Förp.:8
This dirt and stain remover made from 100% melamine resin can be used for removing tough marks, stains and ground-in dirt.
- More efficient than any microfibre cleaning cloth
- No chemicals needed, works with water only