Orders LinkContactUsLinkComponent
His buffer kit
His buffer kit
His Buffer Kit, 1 KIT
Katalog # 11-0034-00
Leverantör:   Cytiva
His buffer kit
Katalog # 11-0034-00
Leverantör:  Cytiva
CAS-nummer:  
His buffer kit
His buffer kit
His Buffer Kit, 1 KIT
CAS-nummer:  
Price
1 800,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 KIT
Product Details & Documents
His Buffer Kit contains carefully prepared buffer concentrates for binding, washing, and elution of histidine-tagged proteins according to recommended protocols. Moreover, a method for optimization of imidazole concentration in the binding buffer is described.
  • Eliminates time-consuming buffer preparation
His Buffer Kit can be used in combination with a number of column types, such as His SpinTrap, His MultiTrap FF, His MultiTrap HP, His GraviTrap, HisTrap FF, HisTrap FF crude and HisTrap HP
Leveransinformation: Kit contains 2×100 ml phosphate 8X stock solution (pH 7,4) and 100 ml highly pure 2 M imidazole (pH 7,4).
Varning: Storage: 4 to 30 °C.
Specifikationer
Specifikationer
  • Förp.:
    1 KIT
  • Beskrivning:
    His Buffer Kit
Specifikationer