High visibility softshell jacket with detachable sleeves, Malden 724Z
Supplier: SIOEN
Certificates
About this item
This lightweight modern jacket is made of a 2-layer bonded softshell fabric consisting of 100% polyester stretch and 100% polyester fleece inside. The jacket provides good protection against wind and cold.
- Straight collar, zip closure with long zip puller and chin protection
- One inset chest pocket with zip closure, two inset pockets with zip closure
- Detachable sleeves
- Sleeve narrowing by touch and close fastening
Can be zipped into other jackets for improved protection.
Back length: 75 cm
Weight: 250 g/m²
Certifications: EN 471 (3-2), EN 14058 (1XXXX)
Specifications
- Catalog No:
- 113-7279
- 113-7277
- 113-7273
- 113-7272
- 113-7278
- 113-7282
- 113-7281
- 113-7276
- 113-7275
- 113-7280
- 113-7274
- 113-7283
- Modell:
- Malden 724Z
- Malden 724Z
- Malden 724Z
- Malden 724Z
- Malden 724Z
- Malden 724Z
- Malden 724Z
- Malden 724Z
- Malden 724Z
- Malden 724Z
- Malden 724Z
- Malden 724Z
- Outfit applikationer:
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Risk area:
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Kön:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Stängningsstorlek:
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Tyg:
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- Vikt:
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²
- 250 g/m²