Flame-retardant fleece lining for jackets, Royston 7220
Supplier: SIOEN
Certificates
About this item
Soft and breathable lining made of 100% double-sided polyester fleece with anti-pilling treatment. The lining provides good cold protection and can be zipped into jackets 7218 and 7219.
- Stitched seams
- Round neck, zip closure
- Slit opening to inside pocket of rain jacket
- Can't be worn separately
Back length: 75 cm
Weight: 275 g/m²
Certifications: EN 14116 (Index 1), EN 14058 (1XXXX)
Specifications
- Catalog No:
- 113-7218
- 113-7217
- 113-7220
- 113-7222
- 113-7219
- 113-7221
- Modell:
- Royston 7220
- Royston 7220
- Royston 7220
- Royston 7220
- Royston 7220
- Royston 7220
- Outfit applikationer:
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Kön:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Stängningsstorlek:
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Tyg:
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Vikt:
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²
- 285 g/m²