ContactUsLinkComponent
Desiccators
Desiccators
  VITL326294
 :  VITLAB
undefined
Desiccators
  VITL326294
 :  VITLAB
  • Beskrivning:
    Desiccators, Ø 273 mm, disc Ø 230 mm
  • Seq:
    03
  • Förp.:
    1
Ideal dessicators for use in educational laboratories.
  • Lid made from PC is sealed with a neoprene gasket
  • Lower portion made from PP can be filled with desiccants
  • Materials to be dried can be placed on a perforated disc made of PP