Centrifugal filters, Amicon Ultra-2 ml
Supplier: Merck
Certificates
About this item
Ultra centrifugal filters provide fast sample processing and promote high sample recoveries, even in dilute samples, through ultrafiltration.
- Heat-sealed membrane eliminates adhesives and downstream extractables
- Enables maximum protein recovery without introducing pipetting errors
- Compatible with pH 1 to 9
- Low binding membrane and PP housing for >90% sample recovery
The unique features of the Amicon® Ultra centrifugal filters gives the fastest, most efficient concentration for sensitive downstream applications.
Specifications
- Catalog No:
- MILFUFC201024
- MILFUFC210024
- MILFUFC205024
- MILFUFC203024
- MILFUFC200324
- Filtreringsyta:
- 1 cm²
- 1 cm²
- 1 cm²
- 1 cm²
- 1 cm²
- Max. RCF:
- 4000/7500 ×g
- 4000/7500 ×g
- 4000/7500 ×g
- 4000/7500 ×g
- 4000/7500 ×g
- Dödvolym:
- <5 µl
- <5 µl
- <5 µl
- <5 µl
- <5 µl
- Ø×H:
- 15,9×119 mm
- 15,9×119 mm
- 15,9×119 mm
- 15,9×119 mm
- 15,9×119 mm
- Provvolym:
- max. 2 µl
- max. 2 µl
- max. 2 µl
- max. 2 µl
- max. 2 µl
Specifications
Related Information
Centrifugal Filter Unit
Videos
Merck Protein Buffer Optimization and Sample Concentration.pdf