ContactUsLinkComponent
Sequencing reaction purification, illustra™ AutoScreen 96 Well Plates
Sequencing reaction purification, illustra™ AutoScreen 96 Well Plates
Katalog # 25-9005-98
Leverantör:  Cytiva
CAS-nummer:  
undefined
Sequencing reaction purification, illustra™ AutoScreen 96 Well Plates
Katalog # 25-9005-98
Leverantör:  Cytiva
CAS-nummer:  
Specifikationer
  • Förp.:
    10
  • Beskrivning:
    illustra™ AutoScreen 96A well plate, for use with MegaBASE sequencer
Specifikationer
Om denna produkt
AutoScreen 96 well plate consists of a 96-well filter plate containing DNA grade Sephadex™ G-50 for purification of sequencing reactions prior to analysis on MegaBACE 1000 and can be used for other size exclusion applications.