Hydrophobic interaction chromatography columns, HiTrap™ Octyl FF Sepharose™
Leverantör: Cytiva
Certifikat
Om denna produkt
HiTrap™ Octyl FF are prepacked Octyl Sepharose™ 4 Fast Flow columns for hydrophobic interaction chromatography (HIC).
- Provide convenient and fast small-scale protein purifications
- Flexible design for use with syringes, single laboratory pump, or chromatography system such as ÄKTA design
- High flow rates and high capacities
- Excellent and easy scale-up achieved by connecting several HiTrap™ columns in series or using HiPrep™ columns
The columns are used for small-scale protein purification as well as for screening of binding and elution conditions. Octyl Sepharose™ 4 Fast Flow is a well-established, standard aliphatic HIC medium for capture and intermediate purification.
Specifikationer
- Catalog No:
- 17-5196-01
- 17-1359-01
- Max. tryck:
- 5 bar
- 5 bar
- pH område:
- 3 - 13
- 3 - 13