High molecular weight transfer buffer (5X)
Catalog # 786-423
Supplier: G-Biosciences
CAS Number:
Specifications
- Förp.:1 L
- Beskrivning:High Molecular Weight Transfer Buffer (5X)
Specifications
About this item
High molecular weight transfer buffer (5X) is designed to facilitate the transfer of notoriously difficult high molecular weight proteins (>70 kDa) during Western blotting.