Long johns, flame-retardant, Borger 2689
Supplier: SIOEN
Certificates
About this item
Anti-static and flame-retardant underwear made of soft handle knitted fabric consisting of 60% Protex, 39% Viloft and 1% AST (Jersey).
- Ribbed ankles for extra comfort, front fly
- Inherently flame-retardant
- Knitted fabric has antimicrobial and moisture regulating properties
Weight: 180 g/m²
Certifications: EN ISO 11612 (A1, B1, C1, F1), EN ISO 14116 (Index 3); EN 1149-5
Specifications
- Catalog No:
- 113-7048
- 113-7053
- 113-7052
- 113-7051
- 113-7050
- 113-7049
- Modell:
- Borger 2689
- Borger 2689
- Borger 2689
- Borger 2689
- Borger 2689
- Borger 2689
- Outfit applikationer:
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Risk area:
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Kön:
- För män
- För män
- För män
- För män
- För män
- För män
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Tyg:
- 60% Protex, 39% Viloft and 1% AST (Jersey)
- 60% Protex, 39% Viloft and 1% AST (Jersey)
- 60% Protex, 39% Viloft and 1% AST (Jersey)
- 60% Protex, 39% Viloft and 1% AST (Jersey)
- 60% Protex, 39% Viloft and 1% AST (Jersey)
- 60% Protex, 39% Viloft and 1% AST (Jersey)
- Vikt:
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²
- 180 g/m²