Hand cleansing wipes, pre-saturated, Kresto kwik-wipes®
Catalog # 115-1473
Supplier: Deb-STOKO
Specifications
- Förp.:70 wipes/bucket, 4 buckets/case
- Beskrivning:Kresto kwik-wipes®
- L×B:260×300 mm
Specifications
About this item
These polypropylene wipes are moistened with a cleansing solution that is ideal for fast cleansing and pre-cleansing after contact with heavy-duty contamination, such as waste mineral and lubricating oils, graphite, metallic dust and carbon black.
- Ideal for workplaces where there is no water available for cleaning hands
- Slightly roughened surface provides consistent and reliable cleansing power
- Non soapy wipes help to stop skin swelling
- Slightly acidic pH helps to maintain the natural acidic protective mantle
Caution: *Produkten/produkterna på denna sida ligger utanför tillämpningsområdet för EU -direktivet 2019/904