Sleeve protector, cut resistant, Aracut® Sleeve 14
Katalog # 112-2919
Leverantör: HONEYWELL SAFETY
Specifikationer
- Passform på plagget:Regular
- Outfit applikationer:Mechanical protection
- Kön:Unisex
- Risk area:Heat,Cuts
- Färg:Gul
- Längd:350 mm
- Tyg:Kevlar®
- Förp.:1
Specifikationer
Om denna produkt
This Kevlar® thread knitted sleeve with thumbhole provides protection of the forearm against cuts and punctures.
- Knitting technique helps to obtain a cuff close to the hand, with good dexterity
- Kevlar® fibre offers good cutting resistance and protection against thermal hazards (contact with heat)
- Two layers of Kevlar®: Greater mechanical and thermal performance of the cuff
- Reinforced thumb gives the cuff better support
Gauge: 10
EN 388 (132X), EN 407 (X1XXXX); Cat. II
Certifications: EN 388 (132X), EN 407 (X1XXXX); Cat. II