Latex gloves, Solo 995
112-0766
: MAPA
- Handskstorlek:9
- Förp.:100/box
- Ej handspecifika/handspecifika:Ej handspecifika
- Engångs/Återanvändbar:Engångs
- Godkända för livsmedelskontakt:+
- Latex/latexfri:Latex
- Färg:Blå
- Längd:240 - 260 mm
- Seq:04
- Tjocklek:0,10 mm
- EN standard:EN ISO 374-1 Type C, EN ISO 374-5
- Kragmodell:Med rullad kant
- Yta:Textured fingertips
These natural latex gloves with chlorinated interior and pebbled fingertips provide good protection for light handling of food, the assembly of small, oil- and grease-free parts in the automotive industry and for laboratory applications.
- Non sterile, chlorinated, textured fingertips
- Excellent dexterity thanks to flexibility of natural latex
- Can be worn alone or as an outer-glove
- Easy to put on and take off
- Protection adapted to short duration usage
: EN ISO 374-1 Type C, EN ISO 374-5; Cat. III
Food contact approved (except oily foods with a reduction factor of under 2, beverages, acid food products, dairy products, fruit and vegetables)
Food contact approved (except oily foods with a reduction factor of under 2, beverages, acid food products, dairy products, fruit and vegetables)