Plugs, PE
Catalog # MANA702807
Supplier: MACHEREY-NAGEL
Specifications
- För:Shell vials, N 8
- Färg:Genomskinlig
- Beskrivning:N 8 PE plug
- Seq:01
- Förp.:100
Specifications
About this item
PE plugs with a diaphragm for safe penetration of the needle are used for the shell vials.
- Easy and quick to seal the vial/closure combination