Capillary columns, CP-Sil 5 CB for Sulphur
Katalog # VARICP7529
Leverantör: VARIAN
Specifikationer
- Kolumnformat:Capillary
- Skikttjocklek:4,00 µm
- Ø inr.:0,32 mm
- Längd:30 m
- Phase:CP-Sil 5 CB for Sulfur
- Förp.:1
Specifikationer
Om denna produkt
- Trace analysis of volatile sulfur compounds to C7 mercaptan for higher productivity
- Non-polar phase providing accurate results based on volatility
- High inertness, elutes SO₂ for high quality data and low detection limits