Pipette aid, Micro-Pipex
Catalog # HECH40558001
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- Beskrivning:Pipetteringshjälpmedel, Micro-Pipex, för mikropipetter för engångsbruk, volympipetter (pipetter för röda och vita blodkroppar) och blodsockerpipetter upp till 1 ml
- Förp.:1
Specifications
About this item
Aid for pipetting liquids by using disposable micropipettes with ring mark as well as other pipettes up to 1 ml, e.g. RBC and WBC pipettes.
- Contaminated disposable micropipettes up to 50 µl are released contactless by the integrated ejection
- Autoclavable at +121 °C
- Pipette holder: max. 5 mm Ø
Please note: The pipetting aid may not be used for capillaries end-to-end.