Kiselgel ≥99.995%, sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 41099.KS
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Förp.:1
- Seq:0001
Specifications
About this item
CAS Number:7631-86-9
:
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
Merck Index:14,08493
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1719 °C
Density:2,2 g/cm³ (20 °C)
Storage Temperature:Omgivande
EINECS:231-545-4
:14808-60-7