Aluminium foils, round
Catalog # GERB5712
Supplier: GERBER FUNKE
Specifications
- Ø:130 mm
- Tjocklek:30 µm
- Förp.:1.000
Specifications
About this item
These aluminium foils are used with reference drier RD-8 for determining the moisture content of powdered milk.