Orders LinkContactUsLinkComponent
Hydrometer cylinders
Hydrometer cylinders
500 ml, 73 mm, 351 mm
Katalog # 614-0005
Leverantör:   VITLAB
Hydrometer cylinders
Katalog # 614-0005
Leverantör:  VITLAB
Hydrometer cylinders
Hydrometer cylinders
500 ml, 73 mm, 351 mm
Price
680,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Produktdetaljer och dokument
Cylinder, made of PP, translucent.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Robust and stable
With spout and overflow vessel. This ensures that the hydrometer can be read with a completely filled cylinder.
Specifikationer
  • Kapacitet:
    500 ml
  • Innerhöjd:
    351 mm
  • Innerdiam.:
    73 mm