Deionizers with nylon resin canisters, behropur®
Leverantör: BEHR
Certifikat
Om denna produkt
Mixed bed ion exchanger for small to medium quantities of ultrapure water to the supply from laboratory dishwashers to general care in the laboratory and for smaller consumers in the industry.
- Comfortable and safe
- Directly and without pressure reducer to the raw water network connectable
- Resistant to alternating pressure
- Optimum utilisation of the ion exchanger capacity
The unique nozzle system distributes the raw water over the whole resin bed.
Conductance is controlled directly on the unit (installed measuring equipment) or with remote location meter.
Specifikationer
- Catalog No:
- BEHRB00223471
- BEHRB00223469
- BEHRB00223473
- Modell:
- B22dN
- B10dN
- B45dN
- Volym:
- 2400 L
- 1200 L
- 5500 L
- Max. flöde:
- 500 l/h
- 300 l/h
- 800 l/h
- Ø×H:
- 210×1120 mm
- 210×680 mm
- 260×1250 mm