Vacuum manifold, CHROMABOND® MULTI 96
Katalog # MANA738630.M
Leverantör: MACHEREY-NAGEL
Specifikationer
- Beskrivning:CHROMABOND® MULTI 96 vacuum manifold for 96-well monoblocks with reservoir tank, 3 spacer pairs, vacuum gauge and control valve
- Seq:13
- Förp.:1 KIT
Specifikationer
Om denna produkt
CHROMABOND® MULTI 96 vacuum manifold is designed for applications of CHROMABOND® MULTI 96 SPE plates as well as for CHROMAFIL® MULTI 96 filter plates.
- Suitable for 96-well monoblocks
- Optimum flow rate due to vacuum adjustment by control valve
- Reservoir tank is included and further accessories are available