Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Rör
Centrifugrör
Centrifuge tubes, glass
Centrifuge tubes, glass
Centrifuge tubes, glass
Leverantör:
MURRAY S
Certifikat
Expandera bilden
Centrifuge tubes, glass
Leverantör:
MURRAY S
Om denna produkt
Soda glass.
Conical or cylindrical
With or without graduations and rim
Withstand forces up to 1300×g