Tubing clips
Catalog # 229-0589
Supplier: MURRAY S
Specifications
- Spännvidd:Ø 14 mm
- Förp.:10
Specifications
About this item
Acetal copolymer.
- For rubber and flexible PVC tubing with an external diameter of up to 14 mm
- Hinged side allows it to be clamped around the tubing already connected to the apparatus
- Screw adjustment ensures fine flow control
- Material is easy to clean, sterilisable, and resistant to most acids and alkalis