Sleeves for vacuum filtration
Katalog # KART420
Leverantör: KARTELL
Specifikationer
- Beskrivning:Set of 7, int. Ø 17 to 75 mm, ext. Ø 21 to 86 mm
- Förp.:1
Specifikationer
Om denna produkt
Set of 7 adapters, conical, made of black neoprene, sleeve supports for Büchner funnels and filter flasks.