Orders LinkContactUsLinkComponent
antibiotics_crop_955x590.jpg
Antibiotics & Antimycotics
Antibiotics and Antimycotics are a type of antimicrobial drug used in the treatment and prevention of bacterial infections. They may either kill or inhibit the growth of microorganisms.
Antibiotics & Antimycotics | Avantor
In science, antibiotics and antimycotics are mostly used in cell culture. They are used to select for a specific type of cells, either when cultivating mammal cells to exclude bacteria or when selecting for genetically modified cells which carry a resistance gene.
PanReac AppliChem antibiotics and antimycotics can be used for small assays and larger scale applications. Here, you can find a selection of the most commonly used products from this range.
Ensure reproducibility for your cell culture or microbiology experiments with PanReac AppliChem antibiotics and antimycotics.
List of selected antibiotics:
Product No. Description Target
organism
Mode
of action
Recommended
working
concentration
Stock
solution
A1907
Amphotericin B
fungi, yeast
binds to sterols with planar structure and impairs with membrane permeability
0.25 µg/ml
>3 µg/ml fungicidal
30-40 mg/ml in DMSO
A3784
Blasticidin S hydrochloride
Prokaryotes, Eukaryotes
inhibits protein biosynthesis by preventing the formation of the peptide bond
3-100 µg/ml
50 mg/ml in H2O or buffer store at +4°C - -20°C
A1491
Carbenicillin disodium salt
gram negative germs, enterococci
inhibits cell wall synthesis (transpeptidase) in growing bacteria
20-60 µg/ml
50 mg/ml in H2O/MeOH 1:1 store at -20°C
A0879
Cycloheximide (Actidion)
fungi, eukaryotes
binds to 80 S ribosome in eukaryotic cells; inhibits formation of peptide bond
10 µg/ml
10 mg/ml
A1492
Gentamycin sulfate
gram positive/ negative germs
inhibits protein synthesis by binding to the L6 protein of the 50 S ribosomal subunit
15-50 µg/ml
10-20 mg/ml in water, formamide
A2175
Hygromycin B - Solution
10-400 µg/ml
ca. 41 mg/ml in H2O; store at -20°C
A1493 & A4789
Kanamycin sulfate & Kanamycin Sulfate (Ph. Eur., BP) pure, pharma rade
gram positive/
negative cocci & bacteria
inhibits protein synthesis (translocation)
10-100 µg/ml
10 mg/ml in H2O
store at -20°C
A0890
Polymyxin B sulfate
gram negative bacteria - non- proliferating germs too
interaction with phospholipid components of the bact. cell membrane; changes permeability of the membrane and causes efflux of essential plasma compounds
50 µg/ml
25 mg/ml H2O, MeOH
A1839
Vancomycin hydrochloride
bacteriostatic and bacterizide against gram positive cocci and bacteria
amphoteric glyopeptide antibiotic; binds to bacterial cell wall precursors (peptidoglycans)
1-25 µg/ml
soluble in water ›100 mg/ml