0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
GENE KNOCKOUT KIT HUMAN BACH1 1.5NMOL
MSPPENSG0000015627
:
MARKET SOURCE PART PROCESS
GENE KNOCKOUT KIT HUMAN BACH1 1.5NMOL
MSPPENSG0000015627
:
MARKET SOURCE PART PROCESS