Order Entry
Sweden
ContactUsLinkComponent
Muffle Furnace L5/11/C550 Tmax1100 with flap door and protective gas connection, over-temperature limiter and thermocouple port in t he rear wall 1 * 1 items
  455-2634
New Product
Muffle Furnace L5/11/C550 Tmax1100 with flap door and protective gas connection, over-temperature limiter and thermocouple port in t he rear wall 1 * 1 items
  455-2634